Learn More
National Diagnostics SequaGel UreaGel 29:1 Concentrate | 1L
SDP
Catalog No. NC1476480
Encompass
UreaGel 29:1 Concentrate is one of three components of the UreaGel 29:1 System. The UreaGel 29:1 System consists of UreaGel 29:1 Concentrate, UreaGel Diluent, and UreaGel Buffer. This system provides a convenient, dependable means for researchers to prepare 29:1 gels of varying percentage. With the UreaGel 29:1 System the researcher readily prepares any commonly used gel formulation up to 20% monomer (29:1 acrylamide:bisacrylamide). Liter bottles of UreaGel Concentrate contain 241.6 grams of acrylamide, 8.3 grams of methylene bisacrylamide, and 7.5M urea in a deionized aqueous solution.
Catalog No. NC1476480 Supplier National Diagnostics Supplier No. EC828
UreaGel 29:1 Concentrate is one of three components of the UreaGel 29:1 System. The UreaGel 29:1 System consists of UreaGel 29:1 Concentrate, UreaGel Diluent, and UreaGel Buffer. This system provides a convenient, dependable means for researchers to prepare 29:1 gels of varying percentage. With the UreaGel 29:1 System the researcher readily prepares any commonly used gel formulation up to 20% monomer (29:1 acrylamide:bisacrylamide). Liter bottles of UreaGel Concentrate contain 241.6 grams of acrylamide, 8.3 grams of methylene bisacrylamide, and 7.5M urea in a deionized aqueous solution.