Learn More
Sigma Aldrich Fine Chemicals Biosciences sequence 9-deoxyribonucleotide
Catalog No. 502901806
Encompass
Random Nonamers are random sequences of nine deoxyribonucleotides (9-mers) used to prime mRNA with or without a poly(A) tail. Random nonamers may be used as universal non-specific primers.
Catalog No. 50-290-1806 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R7647100UL
Random Nonamers are random sequences of nine deoxyribonucleotides (9-mers) used to prime mRNA with or without a poly(A) tail. Random nonamers may be used as universal non-specific primers.