Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Sequencing the Human Microbiome
SDPSCI_ED

Catalog No. s100488 Shop All EDVOTEK Products
Change view
Click to view available options
Quantity:
10 Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S100488 10 Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S100488 Supplier EDVOTEK™ Supplier No. 339340
Only null left
Add to Cart
Add to Cart

In this experiment, students will read DNA sequences obtained from automated DNA sequencing techniques. The data will be analyzed using publicly available databases to identify the bacterial species present in a patient sample. The results will be used to make a diagnosis.

  • Group Size: Sequences for 10 groups
  • Time Required: Complete in 45 minutes
  • Kit Includes: Instructions. This experiment contains a total of twelve sections of automated DNA sequence printouts. Students can use any DNA sequence database to perform the activities in this lab. For purposes of simplification we have chosen to illustrate the database offered by the NCBI.
  • You need: Computer with internet access

Specifications

Format Kit
Sequencing Type Genome and DNA Sequencing
For Use With (Application) Students will read DNA sequences obtained from automated DNA sequencing techniques
Includes Twelve sections of automated DNA sequence printouts; Instructions
Product Type Educational Experiment
Quantity 10 Groups
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.