Learn More
Medchemexpress LLC Sequoyitol (5-O-Methyl-myo-inositol) | 523-92-2 | 5 MG
SDP
Catalog No. 5000420795
Encompass_Preferred
Sequoyitol (5-O-Methyl-myo-inositol) is isolated from plants. It decreases blood glucose, improves glucose intolerance, and is used to treat diabetes. This product is for research use only and not sold to patients.
  • Isolated from plants
  • Decreases blood glucose
  • Improves glucose intolerance
  • Used to treat diabetes
  • For research use only
  • Store at 4°C, protected from light
  • When in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month, protected from light.
Catalog No. 50-004-20795 Supplier Medchemexpress LLC Supplier No. HYN24215MG
Sequoyitol (5-O-Methyl-myo-inositol) is isolated from plants. It decreases blood glucose, improves glucose intolerance, and is used to treat diabetes. This product is for research use only and not sold to patients.
  • Isolated from plants
  • Decreases blood glucose
  • Improves glucose intolerance
  • Used to treat diabetes
  • For research use only
  • Store at 4°C, protected from light
  • When in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month, protected from light.