Learn More
Medchemexpress LLC Ser-(2,3-Bispalmitoyloxypropyl)-Cys-Gly-Asp-Pro-Lys-His-Pro-Lys-Ser-Phe | 322455-70-9 | 99.2% | 1666.16 | 1 MG
SDP
Catalog No. 5000423518
Encompass_Preferred
FSL-1 is a bacterial-derived toll-like receptor 2/6 (TLR2/6) agonist that enhances resistance to experimental HSV-2 infection. It significantly reduces HSV-2 replication in vitro and in human vaginal epithelial cells, and induces resistance to experimental genital HSV-2 infection via a specific cytokine response. FSL-1 also enhances phagocytosis of bacteria by macrophages through a Toll-like receptor 2-mediated signaling pathway.
  • Enhances resistance to experimental HSV-2 infection.
  • Reduces HSV-2 replication in vitro and in human vaginal epithelial cells.
  • Induces resistance to genital HSV-2 infection through specific cytokine response.
  • Enhances phagocytosis of bacteria by macrophages.
  • Functions as a TLR2/6 agonist.
Catalog No. 50-004-23518 Supplier Medchemexpress LLC Supplier No. HYP20361MG
FSL-1 is a bacterial-derived toll-like receptor 2/6 (TLR2/6) agonist that enhances resistance to experimental HSV-2 infection. It significantly reduces HSV-2 replication in vitro and in human vaginal epithelial cells, and induces resistance to experimental genital HSV-2 infection via a specific cytokine response. FSL-1 also enhances phagocytosis of bacteria by macrophages through a Toll-like receptor 2-mediated signaling pathway.
  • Enhances resistance to experimental HSV-2 infection.
  • Reduces HSV-2 replication in vitro and in human vaginal epithelial cells.
  • Induces resistance to genital HSV-2 infection through specific cytokine response.
  • Enhances phagocytosis of bacteria by macrophages.
  • Functions as a TLR2/6 agonist.