Learn More
Medchemexpress LLC (Ser(Ac)3)-Ghrelin (mouse, rat) | 321974-76-9 | 99.55% | 3230.64 | 10 MG
SDP
Catalog No. 5000397547
Encompass_Preferred
(Ser(Ac)3)-Ghrelin (mouse, rat) is a polypeptide identified through peptide screening. Peptide screening serves as a research tool that primarily groups active peptides using immunoassay. This method is applicable for investigating protein interactions, conducting functional analyses, and performing epitope screenings, particularly within the field of agent research and development.
  • Suitable for protein interaction, functional analysis, and epitope screening
  • High purity for reliable research outcomes
  • Polypeptide structure
Catalog No. 50-003-97547 Supplier Medchemexpress LLC Supplier No. HYP468710MG
(Ser(Ac)3)-Ghrelin (mouse, rat) is a polypeptide identified through peptide screening. Peptide screening serves as a research tool that primarily groups active peptides using immunoassay. This method is applicable for investigating protein interactions, conducting functional analyses, and performing epitope screenings, particularly within the field of agent research and development.
  • Suitable for protein interaction, functional analysis, and epitope screening
  • High purity for reliable research outcomes
  • Polypeptide structure