Learn More
Bachem (Ser(PO3H2)202,Thr(PO3H2)205)-Tau Peptide (194-213) Trifluoroacetate
SDP
Supplier:  Bachem 4099870.1000
Encompass_Preferred
(Ser(PO3H2)202,Thr(PO3H2)205)-Tau Peptide (194-213) Trifluoroacetate, H-Arg-Ser-Gly-Tyr-Ser-Ser-Pro-Gly-Ser(PO3H2)-Pro-Gly-Thr(PO3H2)-Pro-Gly-Ser-Arg-Ser-Arg-Thr-Pro-OH, 2022956-55-2, C81H133N29O36P2, Mr 2151.07, 1mg.
Catalog No. 50-194-6633