Learn More
Bachem H-Arg-Ser-Gly-Tyr-Ser-Ser-Pro-Gly-Ser(PO3H2)-Pro-Gly-Thr(PO3H2)-Pro-Gly-Ser-Arg-Ser-Arg-Thr-Pro-OH
SDP
Supplier:  Bachem 4099870.0500
Encompass_Preferred
(Ser(PO3H2)202,Thr(PO3H2)205)-Tau Peptide (194-213) Trifluoroacetate, H-Arg-Ser-Gly-Tyr-Ser-Ser-Pro-Gly-Ser(PO3H2)-Pro-Gly-Thr(PO3H2)-Pro-Gly-Ser-Arg-Ser-Arg-Thr-Pro-OH, 2022956-55-2, C81H133N29O36P2, Mr 2151.07, 0.5mg.
Catalog No. 50-194-6632