Learn More
CPC Scientific H-Ser-Tyr-Leu-Gln-Asp-Ser-Val-Pro-Asp-Ser-Phe-Gln-Asp-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CANS008A
Encompass
ONE-LETTER SEQUENCE: SYLQDSVPDSFQDMOLECULAR FORMULA: C65H93N15O26MOLECULAR WEIGHT:1500.55STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [320341-56-8], SYNONYMS: (Val438)-Tyrosinase (432-444) (human)RESEARCH AREA: Cancer
Catalog No. 50-210-8910