Learn More
Velp™ SER148/3 Solvent Extractor

Description
- Reliable solid-liquid solvent extraction system with three positions
- Rapid analysis with maximum precision or accuracy
- Maximum reproducibility and high flexibility
- Extraction according to Randall method, faster than traditional Soxhlet
- Accepts majority of solvents
- High solvent recovery and low solvent consumption
Specifications
Specifications
| Product Type | Solvent Extractor |
| For Use With (Equipment) | SER 148/3 Solvent Extractor |
| Capacity (Metric) | 150 mL |
| Frequency | 50/60 Hz |
| Voltage | 115 V |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.