Learn More
Velp™ SER148/6 Solvent Extractor
SDP
Catalog No. 11392214
Change view
Click to view available options
Voltage:
115 V
Catalog No. Voltage
11-392-214 115 V
1 options
This item is not returnable. View return policy
Catalog No. 11-392-214 Supplier Velp™ Supplier No. F30310242
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
This six position solvent extractor is a safe and convenient semi-automated extraction system suitable for fat extraction and sample preparation in a wide range of applications in food, feed and other industrial segments.
  • Reliable solid-liquid solvent extraction system with six positions
  • Rapid analysis with maximum precision or accuracy
  • Maximum reproducibility and high flexibility
  • Extraction according to Randall method, faster than traditional Soxhlet
  • Accepts majority of solvents
  • High solvent recovery and low solvent consumption
Specifications
Product Type Solvent Extractor
For Use With (Equipment) SER 148/6 Solvent Extractor
Capacity (Metric) 150 mL
Frequency 50/60 Hz
Voltage 115 V