Learn More
Cayman Chemical [Ser159]-PKCε (149-164) (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502916189
Encompass_Preferred
A peptide substrate for PKCγ, PKCδ, PKCε, PKCη, and PKCξ; has been used as a target substrate for the quantification of PKCγ, PKCδ, PKCε, PKCη, or PKCξ activity in cell-free or cell-based assays
Catalog No. 50-291-6189 Supplier Cayman Chemical Supplier No. 3750810 MG
A peptide substrate for PKCγ, PKCδ, PKCε, PKCη, and PKCξ; has been used as a target substrate for the quantification of PKCγ, PKCδ, PKCε, PKCη, or PKCξ activity in cell-free or cell-based assays