Learn More
Enzo Life Sciences [Ser25] Protein kinase C (19-31) (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP1110001
Encompass_Preferred
Derived from the PKC pseudosubstrate sequence but with an Ala to Ser-25 substitution which makes it a good substrate (Km=0.2 µM) rather than an inhibitor. The resulting sequence is RFARKGSLRQKNV. Alternative name: [Ser25] PKC (19-31). Purity: ≥97%. Long Term Storage: -20°C.
Catalog No. 50-201-1231