Learn More
Apexbio Technology LLC [Ser25] Protein Kinase C (19-31), 1mg. Cas: 136795-05-6 MFCD: MFCD00076724. PKC substrate
SDP
Supplier:  Apexbio Technology LLC A10331
Encompass_Preferred
Protein Kinase C (19-31), (C67H118N26O17), a peptide with the sequence H-ARG-PHE-ALA-ARG-LYS-GLY-SER-LEU-ARG-GLN-LYS-ASN-VAL-OH, MW= 1559.82. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4673