Learn More
CPC Scientific H-Lys(Biotin)-Arg-Phe-Ala-Arg-Lys-Gly-Ser-Leu-Arg-Gln-Lys-Asn-Val-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PPEP006A
Encompass
ONE-LETTER SEQUENCE: K(Biotin)-RFARKGSLRQKNVMOLECULAR FORMULA: C83H144N30O20S1MOLECULAR WEIGHT:1914.3STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [177966-62-0], RESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0220