Learn More
CPC Scientific H-Arg-Phe-Ala-Arg-Lys-Gly-Ser-Leu-Arg-Gln-Lys-Asn-Val-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PKCP048B
Encompass
High affinity protein kinase C pseudosubstrate.ONE-LETTER SEQUENCE: RFARKGSLRQKNVMOLECULAR FORMULA: C67H118N26O17MOLECULAR WEIGHT:1559.82STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [136795-05-6], RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: C. House et al., Science, 238, 1726 (1987)
Catalog No. 50-211-0202