Learn More
Medchemexpress LLC (Ser8)-GLP-1 (7-36) amide, human | 215777-46-1 | 99.1% | 3313.63 | 25 MG
SDP
Catalog No. 5000398965
Encompass_Preferred
(Ser8)-GLP-1 (7-36) amide, human is a glucagon-like peptide 1 amide, which is derived from glucagonogen as a cleavage product of the GLP-1 (1-36) amide peptide. This compound functions as an entero-insulinotropic hormone, meaning it stimulates the glucose-dependent release of insulin from pancreatic β-cells. Additionally, it influences gastrointestinal motility and secretion. These properties make it relevant for research into metabolic diseases and conditions affecting glucose regulation and digestive processes.
  • Glucagon-like peptide 1 amide derivative.
  • Stimulates glucose-dependent insulin release from pancreatic β-cells.
  • Affects gastrointestinal motility and secretion.
  • High purity (99.10%).
Catalog No. 50-003-98965 Supplier Medchemexpress LLC Supplier No. 25MGHYP3622
(Ser8)-GLP-1 (7-36) amide, human is a glucagon-like peptide 1 amide, which is derived from glucagonogen as a cleavage product of the GLP-1 (1-36) amide peptide. This compound functions as an entero-insulinotropic hormone, meaning it stimulates the glucose-dependent release of insulin from pancreatic β-cells. Additionally, it influences gastrointestinal motility and secretion. These properties make it relevant for research into metabolic diseases and conditions affecting glucose regulation and digestive processes.
  • Glucagon-like peptide 1 amide derivative.
  • Stimulates glucose-dependent insulin release from pancreatic β-cells.
  • Affects gastrointestinal motility and secretion.
  • High purity (99.10%).