Learn More
Medchemexpress LLC (Ser8)-GLP-1 (7-36) amide, human | 215777-46-1 | 99.1% | 3313.63 | 50 MG
SDP
Catalog No. 5000399479
Encompass_Preferred
(Ser8)-GLP-1 (7-36) amide, human is a glucagon-like peptide 1 amide derived from glucagonogen. It is a cleavage product of the GLP-1 (1-36) amide peptide. This compound acts as an entero-insulinotropic hormone, which means it stimulates the release of insulin from pancreatic β-cells in a glucose-dependent manner. Additionally, it influences gastrointestinal motility and secretion. The product is intended for research purposes only and is not for sale to patients.
  • Derived from glucagonogen, a cleavage product of GLP-1 (1-36) amide peptide.
  • Functions as an entero-insulinotropic hormone.
  • Induces glucose-dependent insulin release from pancreatic β-cells.
  • Affects gastrointestinal motility and secretion.
Catalog No. 50-003-99479 Supplier Medchemexpress LLC Supplier No. HYP362250MG
(Ser8)-GLP-1 (7-36) amide, human is a glucagon-like peptide 1 amide derived from glucagonogen. It is a cleavage product of the GLP-1 (1-36) amide peptide. This compound acts as an entero-insulinotropic hormone, which means it stimulates the release of insulin from pancreatic β-cells in a glucose-dependent manner. Additionally, it influences gastrointestinal motility and secretion. The product is intended for research purposes only and is not for sale to patients.
  • Derived from glucagonogen, a cleavage product of GLP-1 (1-36) amide peptide.
  • Functions as an entero-insulinotropic hormone.
  • Induces glucose-dependent insulin release from pancreatic β-cells.
  • Affects gastrointestinal motility and secretion.