Learn More
Sigma Aldrich Fine Chemicals Biosciences Sera MagTM SpeedBeadsTM Bl
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE21152104010350
Encompass
Sera-Mag(TM) SpeedBeads(TM) and Sera-Mag(TM) Carboxylate-Modified Magnetic Particles combine a fast magnetic response time and high binding capacity with a large surface area high sensitivity stability physical integrity and fast reaction kinetics. Covalent coupling of biomolecules such as proteins and nucleic acids to carboxyl groups on the surface is easily accomplished using our standard coupling technology. The core of the particle is made by a free radical emulsion polymerization of styrene and acid monomer. One (Sera-Mag(TM)) or two (Sera-Mag(TM) SpeedBeads(TM)) layers of magnetite are then coated onto this core while the surface is modified to minimize non-specific binding of proteins.
Catalog No. 50-290-8525
Add to Cart
Request a Quote