Learn More
Medchemexpress LLC SERABELISIB 10MG
SDP
Catalog No. NC1783195
Encompass_Preferred
Serabelisib (MLN1117) is a selective p110α inhibitor with an IC50 of 15 nM.Purity 99.66%.CAS 1268454-23-4
Catalog No. NC1783195 Supplier Medchemexpress LLC Supplier No. HY1228510MG
Serabelisib (MLN1117) is a selective p110α inhibitor with an IC50 of 15 nM.Purity 99.66%.CAS 1268454-23-4