Learn More
Aviva Systems Biology Corp SERAC1 Antibody - N-terminal region (ARP60701_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60701_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SERAC1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SERAC1. Alternate names: FLJ14917, FLJ30544ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-132