Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ ACCURUN 1™ Series Blood Donor Multi-Marker Positive Controls

Catalog No. 22554110 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
12 x 3.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-110 12 x 3.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-554-110 Supplier SeraCare™ Supplier No. 20000010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Premier multi-marker controls specifically designed for use in blood donor screening laboratories

  • Formulated to perform within the dynamic range of specific U.S. and internationally available test kits
  • Ready to use and contain markers commonly screened for donated blood
  • Extended open vial stability of up to 60 days
  • Contain HBsAg and antibodies to HIV-1, HIV-2, HCV, HTLV 1, HTLV II, HBc, and CMV
  • In vitro diagnostic use
  • Available in a wide variety of convenient package sizes

  • Combine LabLink xL with the ACCURUN products for an IVD exclusive infectious disease and nucleic acid testing Quality Assurance Program (QAP) from Fisher HealthCare
  • With the LabLink xL QAP program, a lab's performance can be compared to peer laboratories' performances and used to benchmark performance to group standards:
    • Track how other lab's signal to cutoff ratio (s/co) or recovered values (mean values) compare
    • Compare how a lab's variability (standard deviation) compares to a group average
    • Determine if a lab's s/co or recovered values are statistically within acceptable group limits

Specifications

Certifications/Compliance IVD, CE, HC
Content And Storage 2°C to 8°C
For Use With (Equipment) Bio-rad™ genetic system HIV-1/HIV-2 Plus O EIA
Type ACCURUN 1 Multi-analyte Positive Control
Intended Use Detection of Antibodies to HIV 1 and 2, HTLV I and II, HBcAg, HCV, Cytomegalovirus, HBsAg
Matrix Human Serum or Plasma
Quantity 12 x 3.5 mL
Stability 60 Days Open Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.