Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ ACCURUN™ 125 Anti-HBs Positive Control Kit

Catalog No. 22554202 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
6 x 3.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-202 6 x 3.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-554-202 Supplier SeraCare™ Supplier No. 20150078
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For qualitative detection of antibodies to Hepatitis B surface antigen

  • Manufactured from human serum or plasma
  • For use with in-vitro diagnostic test kits
  • For qualitative detection of antibodies to Hepatitis B surface antigen (anti-HBs)
  • 6 x 3.5mL

Specifications

Certifications/Compliance IVD, CE, HC
Type ACCURUN 125 anti-HBs Positive Control
Intended Use Qualitative detection of Antibodies to Hepatitis B Surface Antigen (Anti-HBs)
Matrix Human Serum or Plasma
Quantity 6 x 3.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.