Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ ACCURUN™ 155 Anti-Treponema pallidum (Syphilis) Positive Control

Catalog No. 22554512 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
3 x 2 mL
12 x 3.5 mL
Format:
Tube
Vial
Matrix:
Human Plasma
Human Serum or Plasma
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Matrix
22-554-512 3 x 2 mL Tube Human Serum or Plasma
22-554-185 12 x 3.5 mL Tube Human Plasma
22-554-511 3 x 2 mL Vial Human Serum or Plasma
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-554-512 Supplier SeraCare™ Supplier No. 20150093
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed to closely mimic human samples for accuracy and efficiency

  • Human plasma based controls mimic patient samples
  • Independent controls meet compliance requirements of regulatory agencies and professional organizations
  • Liquid, ready-to-use: no reconstitution or special handling required
  • Extended shelf life means efficiency: allows user to monitor assay performance with same lot over an extended period of time
  • Formulated for use with in vitro diagnostic test kits for the qualitative determination of IgG antibodies to Treponema pallidum (Syphilis), including assay procedures for blood screening, and for the diagnosis and monitoring of patients infected with T. pallidum
  • Manufactured from human serum or plasma, reactive for anti-Treponema

Specifications

Certifications/Compliance IVD, CE, HC
Content And Storage 2°C to 8°C
Format Tube
Type ACCURUN 155 Anti-Treponema Pallidum Positive Control
Intended Use Qualitative determination of IgG Antibodies to Treponema pallidum (Syphilis)
Matrix Human Serum or Plasma
Quantity 3 x 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.