Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ ACCURUN™ 190 Anti-Trypanosoma cruzi (Chagas) Positive Control

Catalog No. 22554513 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
6 x 3.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-513 6 x 3.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-554-513 Supplier SeraCare™ Supplier No. 20150097
Add to Cart
Add to Cart
This item is not returnable. View return policy

Enhances the quality and precision of Chagas test results, day-to-day and run-to-run

  • Human plasma based control mimics patient samples
  • Independent controls meet compliance requirements of regulatory agencies and professional organizations
  • Liquid, ready-to-use: no reconstitution or special handling required
  • Extended shelf life means efficiency: allows user to monitor assay performance with same lot over an extended period of time
  • Manufacturing facilities are ISO 13485 certified and cGMP compliant

Specifications

Certifications/Compliance ISO 13485, CE, IVD
Format Tube
Type ACCURUN 190 Anti-Trypanosoma cruzi Positive Control
Intended Use Qualitative detection of Anti-Trypanosoma cruzi (chagas)
Matrix Human Serum or Plasma
Quantity 6 x 3.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.