Learn More
SeraCare™ ACCURUN™ 632 Cystic Fibrosis 32 Control
Catalog No. 22554476 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
22-554-476 100 μL
1 options
Catalog No. 22-554-476 Supplier SeraCare™ Supplier No. 20400011
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
A synthetic, multiplex control formulated for research use only
  • Covers all 32 mutations in the Abbott/Celera Cystic Fibrosis Genotyping Assay (CF IVD Core OLA)
  • Eliminates errors arising from use of rotating gDNA control schemes with incomplete mutation profiles
  • Controls contain a mixture of oligonucleotides that produce a call for every mutation tested
  • All mutations are contained in a single vial
  • High purity, large lots, and stable materials–no additional processing required
Not for use in diagnostic procedures
Specifications
Certifications/Compliance RUO
Type ACCURUN 632 Cystic Fibrosis 32 Control
Intended Use Detects significant errors with the use of rotating gDNA control schemes, research use only