Learn More
SeraCare™ ACCURUN™ CMV DNA Positive Control
Catalog No. 22554526 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
5 x 0.75 mL
Catalog No. Quantity
22-554-526 5 x 0.75 mL
1 options
Catalog No. 22-554-526 Supplier SeraCare™ Supplier No. A3505334/DEL/DISC
Independent controls for CMV enhance the consistency and precision of test results, day-to-day and run-to-run
  • Human plasma based controls mimic patient samples
  • Independent controls meet compliance requirements of regulatory agencies and professional organizations
  • Liquid, ready-to-use: no reconstitution or special handling required
  • Extended shelf life means efficiency: allows user to monitor assay performance with same lot over an extended period of time
Specifications
Type ACCURUN 350 CMV DNA Positive Control
Intended Use Detect CMV DNA, for research use only
Matrix Defibrinated Plasma
Quantity 5 x 0.75 mL