Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ ACCURUN™ HBV DNA Positive Control

Catalog No. 22554523 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Certifications/Compliance:
IVD, CE
RUO
Quantity:
5 x 0.6 mL
5 x 4 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance Quantity
22-554-523 IVD, CE 5 x 4 mL
22-554-521 RUO 5 x 0.6 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-554-523 Supplier SeraCare™ Supplier No. 20200096
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Independent controls for HIV enhance the consistency and precision of test results, day-to-day and run-to-run

  • Human plasma based controls mimic patient samples
  • Independent controls meet compliance requirements of regulatory agencies and professional organizations
  • Liquid, ready-to-use: no reconstitution or special handling required
  • Extended shelf life means efficiency: allows user to monitor assay performance with same lot over an extended period of time

Specifications

Certifications/Compliance IVD, CE
Format Vial
Type ACCURUN HBV DNA Positive Control
Intended Use Detect Hepatitis B Virus DNA
Matrix Human Serum or Plasma
Quantity 5 x 4 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.