Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ ACCURUN™ HIV RNA Positive Control

Catalog No. 22554144 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
10 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-144 10 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-554-144 Supplier SeraCare™ Supplier No. 20200087
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Independent controls for HIV enhance the consistency and precision of test results, day-to-day and run-to-run

  • Human plasma based controls mimic patient samples
  • Independent controls meet compliance requirements of regulatory agencies and professional organizations
  • Liquid, ready-to-use: no reconstitution or special handling required
  • Extended shelf life offers efficiency: allows user to monitor assay performance with same lot over an extended period of time

Specifications

Certifications/Compliance IVD, CE, HC
Content And Storage -70°C
Format Vial
Type ACCURUN 315 Series 150
Intended Use Detect and quantitate HIV-1 RNA
Quantity 10 x 1.2 mL
Source Human Plasma
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.