Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ ACCURUN™ Independent Positive Controls For Abbott ARCHITECT™

Catalog No. 22554472 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Format:
Tube
Vial
Quantity:
3 x 5 mL
12 x 3.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Quantity
22-554-472 Tube 12 x 3.5 mL
22-554-471 Vial 3 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Handle ACCURUN controls and all human blood products as though capable of transmitting infectious agents. Use the Centers for Disease Control (CDC) recommended universal precautions for handling ACCURUN. Do not pipette by mouth; do not smoke, eat or drink in areas where specimens are being handled. Clean any spillage by immediately wiping up with 0.5% sodium hypochlorite solution. Dispose of all specimens, controls and materials used in testing as though they contain infectious agents.
Catalog No. 22-554-472 Supplier SeraCare™ Supplier No. 20150066
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Handle ACCURUN controls and all human blood products as though capable of transmitting infectious agents. Use the Centers for Disease Control (CDC) recommended universal precautions for handling ACCURUN. Do not pipette by mouth; do not smoke, eat or drink in areas where specimens are being handled. Clean any spillage by immediately wiping up with 0.5% sodium hypochlorite solution. Dispose of all specimens, controls and materials used in testing as though they contain infectious agents.

For use with in vitro diagnostic test methods that detect HIV-1 antigen

ACCURUN 106 Series 1000 is prepared by diluting HIV-1 viral lysate derived from human cell culture in human serum or plasma nonreactive for HBsAg and negative for antibodies to HIV-1 and 2, HCV and HTLV

  • For use with Abbott ARCHITECT
  • For in vitro diagnostic use

  • Helps to enhance quality control and quality assurance in testing labs
  • Human plasma based controls mimic patient samples
  • Independent controls meet compliance requirements of regulatory agencies and professional organizations
  • Liquid, ready-to-use - no reconstitution or special handling required
  • Extended shelf life means efficiency for labs, allowing assay performance to be monitored with same lot over an extended period of time

  • All SeraCare facilities are ISO 9001 and 13485 certified and comply with cGMP standards
  • Routine use of independent controls helps your lab monitor day-to-day test variability, lot-to-lot variability and staff performance

Specifications

Certifications/Compliance ISO 9000 13485, IVD, CE
Content And Storage 2°C to 8°C
Format Tube
For Use With (Equipment) Abbott Architect™
Type ACCURUN 106 HIV-1 Antigen Positive Control
Intended Use Detect HIV-1 Antigen
Matrix Lysate derived from human cell culture in guman serum or plasma
Quantity 12 x 3.5 mL
Stability 60 Days Open Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.