Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ ACCURUN™ Nucleic Acid Multi-Analyte Positive Controls

Catalog No. 22554525 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
5 x 4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-525 5 x 4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

Use the Centers for Disease Control (CDC) recommended universal precautions for handling ACCURUN. Do not pipette by mouth; do not smoke, eat or drink in areas where specimens are being handled. Clean any spillage by immediately wiping up with 0.5% sodium hypochlorite solution. Dispose of all specimens, controls and materials used in testing as though they contain infectious agents.

To prevent formation of potentially explosive compounds due to reactions of sodium azide and copper or lead pipes, flush waste lines with large quantities of water.

Catalog No. 22-554-525 Supplier SeraCare™ Supplier No. 20200105
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use the Centers for Disease Control (CDC) recommended universal precautions for handling ACCURUN. Do not pipette by mouth; do not smoke, eat or drink in areas where specimens are being handled. Clean any spillage by immediately wiping up with 0.5% sodium hypochlorite solution. Dispose of all specimens, controls and materials used in testing as though they contain infectious agents.

To prevent formation of potentially explosive compounds due to reactions of sodium azide and copper or lead pipes, flush waste lines with large quantities of water.

Independent controls for nucleic acid testing

  • An independent, easy-to-use, fully-characterized, multi-analyte positive control for multiplex or standard NAT donor screening or patient monitoring

Formulated for use with in vitro diagnostic test methods that detect HIV-1 RNA, HCV RNA and HBV DNA

  • ACCURUN 345 Series 150 is prepared by diluting separate stocks of known titers of HIV-1 RNA, HCV RNA and HBV DNA with defibrinated plasma
  • Formulated to low concentration levels of HIV-1 RNA, HCV RNA and HBV DNA; challenge assay performance near limit of detection
  • Specifically formulated for manual and automated assay platforms to help monitor both assay and instrument integrity
  • Human plasma based controls mimic patient samples
  • Monitor all aspects of sample testing, from sample lysis and extraction through PCR amplifiction and detection
  • Independent controls meet compliance requirements of regulatory agencies and professional organizations
  • Liquid, ready-to-use - no reconstitution or special handling required
  • Extended shelf life means efficiency for labs, allowing assay performance to be monitored with same lot over an extended period of time

  • All SeraCare facilities are ISO 9001 and 13485 certified and comply with cGMP standards
  • Routine use of independent controls helps monitor day-to-day test variability, lot-to-lot variability and staff performance

Specifications

Certifications/Compliance IVD, CE, HC
Detectable Analytes HIV-1 RNA, HCV RNA, HBV DNA
Format Vial
Type ACCURUN 345 Multi-analyte Positive Control
Intended Use Detect HIV-1 RNA, HCV RNA and HBV DNA, for research use only
Matrix Defibrinated Plasma
Quantity 5 x 4 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.