Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ ACCURUN™ QC Blood Donor NAT Screening Controls

Catalog No. 22554129 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
10 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-129 10 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-554-129 Supplier SeraCare™ Supplier No. 20350007
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quality control materials for use in blood donor screening

  • Meet the demands of the need for greater sensitivity, complexity, and effective quality control
  • Help blood bankers monitor highly complex assays and fulfill regulatory requirements

  • Combine LabLink xL with the ACCURUN products for an IVD exclusive infectious disease and nucleic acid testing Quality Assurance Program (QAP) from Fisher HealthCare
  • With the LabLink xL QAP program, a lab's performance can be compared to peer laboratories' performances and used to benchmark performance to group standards:
    • Track how other lab's signal to cutoff ratio (s/co) or recovered values (mean values) compare
    • Compare how a lab's variability (standard deviation) compares to a group average
    • Determine if a lab's s/co or recovered values are statistically within acceptable group limits

Specifications

Certifications/Compliance IVD, CE, HC
Content And Storage -20°C
Detectable Analytes HIV-1 RNA, HCV RNA, HBV DNA
Type ACCURUN 803 Multi-analyte Nucleic Acid Negative Control
Intended Use Detect HIV-1 RNA, HCV RNA, HBV DNA, Blood donor screening
Matrix Human Serum or Plasma
Quantity 10 x 1.2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.