Learn More
SeraCare™ AccuSet™ Anti-HBc IgM Mixed Titer Performance Panel Modified
Catalog No. 22554006 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
22 x 1.0 mL
Catalog No. Quantity
22-554-006 22 x 1.0 mL
1 options
This item is not returnable. View return policy
Catalog No. 22-554-006 Supplier SeraCare™ Supplier No. 08050216
This item is not returnable. View return policy
Demonstrates anti-HBc IgM reactivity ranging from negative to strong positive
  • Includes 23 member, 1.0 mL per vial panel of undiluted, naturally-occurring plasma samples
  • Includes test results from commercially-available HBV assays for characterization of panel member
  • Preservatives-free
Specifications
Description AccuSet™ Anti-HBc IgM Mixed Titer Performance Panel Modified
Includes 22 member panel of undiluted, naturally-occurring plasma samples
Quantity 22 x 1.0 mL
For RUO only