Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ AccuSet™ Dengue Early Infection Performance Panel

Catalog No. 22554007 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
0.5 mL/Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-007 0.5 mL/Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-554-007 Supplier SeraCare™ Supplier No. 08450050

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

To develop, evaluate or troubleshoot Dengue test methods

Six-member panel of undiluted, naturally occurring plasma samples, collected in 2010 from a 43-year old male during the development of a Dengue infection and subsequent response. No preservatives added.

  • Store frozen at -70°C or colder to preserve Dengue RNA
  • Dengue antigen and antibody preserved at -20°C or colder
  • 0.5mL/Vial

Specifications

Type Dengue Early Infection Performance Panel
Quantity 0.5 mL/Vial

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.