Learn More
SeraCare™ AccuSet™ HBsAg Mixed Titer Performance Panel Modified
Catalog No. 22554005 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
20 x 1.5 mL
Catalog No. Quantity
22-554-005 20 x 1.5 mL
1 options
This item is not returnable. View return policy
Includes potentially infectious materials
Catalog No. 22-554-005 Supplier SeraCare™ Supplier No. 08050217
This item is not returnable. View return policy
Includes potentially infectious materials
Provides broad range of reactivity for HBsAg
  • Includes 20 member, 1.5 mL per vial panel of undiluted, naturally-occurring plasma samples
  • Includes test results from commercially-available HBV assays for characterization of panel members
  • Preservatives-free
Specifications
Quantity 20 x 1.5 mL
For RUO only