Learn More
SeraCare™ AccuSet™ HIV p24 Antigen Mixed Titer Performance Panels Modified
Catalog No. 22554003 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
18 Member
8 Member
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-003 8 Member
22-554-002 18 Member
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Includes potentially infectious materials.
Catalog No. 22-554-003 Supplier SeraCare™ Supplier No. 08000302
This item is not returnable. View return policy
Includes potentially infectious materials.
For use by manufacturers and laboratories to evaluate the sensitivity and specificity of HIV antigen and HIV antigen/antibody combination assays using well-characterized specimens
  • Includes 18 member, 1.5mL per vial panel of undiluted and naturally-occurring plasma samples collected from asymptomatic blood donors from 1997 through 2008
  • Originated from HIV p24 Mixed Titer Performance Panel PRA204-1.5
  • Includes Fiebig stage and test results for 13 commercially available assays for HIV p24 antigen, HIV antigen/antibody, anti-HIV, and HIV RNA
  • HIV p24 antigen concentration ranges from <3 to >400 pg/mL in these samples
  • Represents Fiebig stages II through VI
  • Preservatives free
Specifications
Type HIV p24 Antigen Mixed Titer
Quantity 8 Member
For Research Use Only (RUO)