Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ AccuSet™ Syphilis Mixed Titer Performance Panel Modified

Catalog No. 22554661 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
17 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-661 17 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
For Research use only
Catalog No. 22-554-661 Supplier SeraCare™ Supplier No. 08200191

missing translation for 'validMainframeMsg'

For Research use only

Intended to help develop, evaluate or troubleshoot syphilis test methods. Syphilis Mixed Titer Performance Panel Modified is a 17-member panel of undiluted, naturally occurring plasma samples. Panel members have reactivity ranging from negative to highly positive for treponema antibody and reagin.

  • Characterized samples and comprehensive data are provided for comparative analysis.
  • No preservatives were added
  • 1 vial per member, 17 members, 0.5mL per vial

Specifications

Content And Storage ≤-20°C
Quantity 17 x 0.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.