Learn More
SeraCare™ AccuSpan™ CMV Linearity Panel
Catalog No. 22554663 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
9 x 2.0 mL
Catalog No. Quantity
22-554-663 9 x 2.0 mL
1 options
Catalog No. 22-554-663 Supplier SeraCare™ Supplier No. 24100174
Specifications
Includes 9-member panel: 7-members positive for CMV DNA representing serial log dilutions in negative diluent; One negative member; One negative diluent member
Quantity 9 x 2.0 mL