Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ AccuVert™ HAV Serconversation Panels

Catalog No. 22554000 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
8 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-000 8 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Includes potentially infectious materials
Catalog No. 22-554-000 Supplier SeraCare™ Supplier No. 06150026
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Includes potentially infectious materials

Helps to determine change in expression from negative to positive of HAV antibody

  • Includes 8-member panel of undiluted, naturally occurring plasma samples
  • Represents serial bleeds collected from a single patient ove course of 27 days during development of a Hepatitis A (HAV) infection and subsequent response
  • Includes test results from commercially-available HAV antibody assays for characterization of panel members
  • Preservatives-free
  • One vial of 1.2mL per member

Specifications

Quantity 8 x 1.2 mL

For RUO only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.