Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ Anti-EBV Mixed Titer Performance Panels

Catalog No. 22554539 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
25 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-539 25 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
These materials have not been inactivated and should be handled, like all blood products, as if potentially biohazardous following good laboratory safety practice. Never pipette by mouth. Do not smoke, eat or drink in areas where specimens are handled. These materials should be disposed of in a manner that will inactivate pathogenic agents.
Catalog No. 22-554-539 Supplier SeraCare™ Supplier No. 08500023

missing translation for 'validMainframeMsg'

These materials have not been inactivated and should be handled, like all blood products, as if potentially biohazardous following good laboratory safety practice. Never pipette by mouth. Do not smoke, eat or drink in areas where specimens are handled. These materials should be disposed of in a manner that will inactivate pathogenic agents.

Used in the evaluation of infectious mononucleosis heterophile and Epstein-Barr virus specific antibody tests

Samples have been selected to demonstrate IgG and/or IgM reactivity; in addition, one negative serum and three negative plasmas have been included as nonreactive controls

  • For research use only

  • Extensive array of panels helps labs maintain the highest standards for assay validation, regulatory compliance and method development
  • Each panel includes comprehensive, comparative data from a variety of methods and commercially available test kits, allowing reagent assay/platform performance, sensitivity and specificity to be independently challenged
  • Assembled from highly characterized, disease state patient samples, and developed specifically for infectious disease testing
  • Enables labs to evaluate, monitor and troubleshoot test and staff performance
  • Minimizes down time and accelerates validation and verification of new test methods, reagent systems and platforms

  • SeraCare panels are rigorously tested and characterized using the highest quality test methods
  • Comprehensive test results provided with each panel; quantified against international standards when available
  • Long-term stability and availability
  • All SeraCare facilities are ISO 9001 and 13485 certified and comply with cGMP standards

Specifications

Certifications/Compliance ISO 9000 13485
Content And Storage Frozen
Format Panel
Type Anti-EBV Mixed Titer Performance Panel
Includes Set of 25 members: 21 members with Infectious Mononucleosis heterophile antibody and/or EBV specific reactivity, one negative Serum and three negative Plasmas as nonreactive controls
Intended Use Evaluating infectious Mononucleosis heterophile and Epstein Barr Virus specific Antibody tests with characterized samples, to provide comprehensive data for comparative analysis
Matrix Serum and Plasma
Quantity 25 x 0.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.