Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ Anti-HBc IgM Mixed Titer Performance Panel

Catalog No. 22554463 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
25 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-463 25 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-554-463 Supplier SeraCare™ Supplier No. 08050192

Package of 25 members with reactivities ranging from negative to strongly positive for anti-HBc IgM

  • Undiluted samples collected from multiple donors of varied analyte concentration, genotype and ethnic/demographic diversity
  • Maintain the standards for assay validation, regulatory compliance and method development
  • Includes comprehensive, comparative data from a variety of methods and commercially available test kits, allowing user to independently challenge reagent assay/platform performance, sensitivity and specificity
  • Assembled from highly characterized, disease state patient samples, and developed specifically for infectious disease testing, both serology and NAT
  • Evaluate, monitor, and troubleshoot test and staff performance
  • Minimize down time and accelerates validation and verification of new test methods, reagent systems and platforms
  • For research use only

Specifications

Content And Storage -10°C
Format Panel
Type Anti-HBc IgM Mixed Titer Performance Panel
Includes Twenty-five Plasma samples with reactivities ranging from negative to strongly positive for Anti-HBc IgM
Intended Use Used to challenge their Anti-HBc IgM test system with regard to sensitivity, specificity, reproducibility, and lot-to-lot variability, to train staff and assess competence
Quantity 25 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.