Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ Anti-HIV 1/2 AccuTrak™ Verification Panel

Catalog No. 22554605 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
5 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-605 5 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-554-605 Supplier SeraCare™ Supplier No. 24050046

Consists of five members with reactivity in anti-HIV 1/2 rapid assays

  • Positive panel members are naturally occurring, undiluted human plasma samples
  • Three members have various reactivities for anti-HIV-1, one member is reactive for anti-HIV-2, and one member non-reactive for anti-HIV 1/2
  • Fill size: one vial per member, five members, 0.5mL per vial
  • Panel members were filtered through 0.2 micron filter.
  • ProClin™ (0.1%) added as preservative

Specifications

Type Anti-HIV 1/2 AccuTrak Verification Panel
Quantity 5 x 0.5 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.