Learn More
SeraCare™ Anti-HIV-1/2 Verification Panel
Catalog No. 22554595 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
5 x 0.5 mL
Catalog No. Quantity
22-554-595 5 x 0.5 mL
1 options
This item is not returnable. View return policy
Catalog No. 22-554-595 Supplier SeraCare™ Supplier No. 24050035
This item is not returnable. View return policy
Five plasma specimens with established reactivity in anti-HIV 1/2 rapid assays
  • May be used as part of an ongoing program for lot acceptance, internal proficiency testing, isolation of system errors and troubleshooting assays, and as part of an ongoing laboratory quality assurance program
  • Positive panel members are naturally occurring, undiluted human plasma samples
  • Three members have various reactivities for anti-HIV-1, one member is reactive for anti-HIV-2, and one member nonreactive for anti-HIV 1/2
  • The non-reactive member is formulated from anti-HIV 1/2 negative pools
  • Panel members were filtered through a 0.2 micron filter; ProClin™ (0.1%) preservative added
Specifications
Format Panel
Type Anti-HIV 1/2 Verification Panel
Includes Five Plasma specimens: Three members have various reactivities for Anti-HIV-1, one member is reactive for Anti-HIV-2, one member is nonreactive for Anti-HIV 1/2
Intended Use Used as part of an ongoing program for lot acceptance, internal proficiency testing, isolation of system errors, troubleshooting assays, laboratory quality assurance program
Quantity 5 x 0.5 mL