Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ Anti-HIV-1 Low Titer Performance Panels

Catalog No. 22554455 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
20 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-455 20 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
These materials have not been inactivated and should be considered biohazardous. Panel members were tested and most were found positive by tests for anti-HIV 1. The units that make up this panel were tested for HBsAg and anti-HCV. All panel members were found to be non-reactive for HBsAg and anti-HCV. Do not pipette by mouth.
Catalog No. 22-554-455 Supplier SeraCare™ Supplier No. 08000241

missing translation for 'validMainframeMsg'

These materials have not been inactivated and should be considered biohazardous. Panel members were tested and most were found positive by tests for anti-HIV 1. The units that make up this panel were tested for HBsAg and anti-HCV. All panel members were found to be non-reactive for HBsAg and anti-HCV. Do not pipette by mouth.

For the evaluation of HIV test systems

Panel of undiluted plasma samples collected from multiple donors; includes members with anti-HIV-1 reactivity near the sensitivity limit and one member negative for anti-HIV-1, included as a nonreactive control

  • For research use only

  • Extensive array of panels helps labs maintain the highest standards for assay validation, regulatory compliance and method development
  • Each panel includes comprehensive, comparative data from a variety of methods and commercially available test kits, allowing reagent assay/platform performance, sensitivity and specificity to be independently challenged
  • Assembled from highly characterized, disease state patient samples, and developed specifically for infectious disease testing
  • Enables labs to evaluate, monitor and troubleshoot test and staff performance
  • Minimizes down time and accelerates validation and verification of new test methods, reagent systems and platforms

  • Rigorously tested and characterized using highest-quality test methods
  • Comprehensive test results provided with each panel; quantified against international standards when available
  • Long-term stability and availability
  • SeraCare facilities are ISO 9001 and 13485 certified and comply with cGMP standards

Specifications

Content And Storage -10°C
Format Panel
Type Anti-HIV-1 low Titer Performance Panel
Includes Set of 20 aliquots from Plasma units: 19 members have Anti-HIV-1 reactivity near the sensitivity limits of Anti-HIV screening tests, one specimen negative for Anti-HIV-1 as non reactive control
Intended Use Evaluating HIV test system using well characterized specimens, to provide comprehensive data for comparative analysis
Quantity 20 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.