Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ Anti-HTLV I/II Mixed Titer Performance Panel

Catalog No. 22554801 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
21 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-801 21 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-554-801 Supplier SeraCare™ Supplier No. 08200178

missing translation for 'validMainframeMsg'

Undiluted, naturally occurring plasma samples collected between 1998 and 2011 from different individuals

  • Ten samples are confirmed positive for anti-HTLV-I by Western blot; Ten are confirmed positive for anti-HTLV-II; One sample is negative for anti-HTLV by all methods used
  • All members were processed by filtration through 0.8μm filters
  • No preservatives
  • For research use only; Not for use in diagnostic procedures

Specifications

Content And Storage -10°C
Format Panel
Type Anti-HTLV I/II Mixed Titer Performance Panel
Includes Set of 21 undiluted aliquots from Plasma units: 10 members positive for Anti-HTLV I, 10 members positive for Anti-HTLV II, one member is non-reactive
Intended Use Evaluating Anti-HTLV I, Anti-HTLV II, Anti-HTLV I/II test system using well characterized specimens, to provide comprehensive data for comparative analysis
Quantity 21 x 1.2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.