Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ Anti-HTLV I/II Qualification Panel

Catalog No. 22554203 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Source:
Human Serum or Plasma
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Source
22-554-203 Human Serum or Plasma
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-554-203 Supplier SeraCare™ Supplier No. 24000181
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Six member panel of samples with established reactivity in anti-Human T-Lymphotropic Virus Types I and/or II

  • Manufactured from human serum or plasma
  • Store at 2° to 8°C
  • For research use only
  • 1 vial per member, 6 members, 3.5mL per vial

Specifications

Content And Storage 2°C to 8°C
Sample Type Six Member Panel of Samples
Type Anti-HTLV I/II Qualification Panel
Source Human Serum or Plasma
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.