Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ Anti-T. cruzi (Chagas) Performance Panel

Catalog No. 22554644 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
21 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-644 21 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-554-644 Supplier SeraCare™ Supplier No. 08450119

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Naturally occurring plasma specimens with reactivity ranging from negative to strongly positive for anti-T.cruzi

  • Specimens are undiluted aliquots from plasma units collected from blood donors in the United States, Central and South America and Mexico
  • All panel members were found nonreactive for HBsAg, anti-HIV and anti-HCV
  • For research use only: Not for use in diagnostic procedures

Specifications

Content And Storage -20°C
Format Panel
Type Anti-T. cruzi (Chagas) Performance Panel
Includes Set of 21 aliquots from Plasma units with reactivity ranging from negative to strongly positive for Anti-T.cruzi
Intended Use Evaluating chagas test system using well characterized specimens, to provide comprehensive data for comparative analysis
Quantity 21 x 1.2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.