Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ BaseMatrix™ Diluent

Catalog No. 22554250 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
12 x 3.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-250 12 x 3.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-554-250 Supplier SeraCare™ Supplier No. 18050074
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cost-effective human serum substitute, a preferred choice for use as diluent

Basematrix diluent is formulated for use with in vitro diagnostic test kits for the detection of HBV DNA, HCV RNA, HIV-1 RNA, Human Parvovirus B19 DNA, HAV RNA, HBsAg, anti-HIV 1/2, anti-HTLV I/II, anti-HBc, and anti-HCV. The product is manufactured from dialyzed, defibrinated human plasma that is negative for HBV DNA, HCV RNA, HIV-1 RNA, Human Parvovirus B19 DNA, and HAV RNA, and non-reactive for HBsAg and anti-HIV-1/2, anti-HCV, anti-HTLV I/II, anti-HBs, and anti-HBc. Basematrix Diluent canbe used as a sample diluent for titration studies. For laboratory use only.

  • Visually clear, free of precipitate, virtually free of particulate
  • Low bio-burden: routinely <10 colony forming units (CFU) per mL
  • Salt balanced close to physiological conditions

Specifications

Content And Storage -20°C
Type Basematrix Diluent
Intended Use Detection of HBV DNA, HCV RNA, HIV-1 RNA, Human Parvovirus B19 DNA, HAV RNA, HBsAg, Anti-HIV 1/2, Anti-HTLV I/II, Anti-HBc, Anti-HCV
Matrix Human Plasma
Quantity 12 x 3.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.