Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ HBsAg Sensitivity Panels

Catalog No. 22554284 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
25 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-284 25 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-554-284 Supplier SeraCare™ Supplier No. 08200184
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Content And Storage -20°C
Type Anti-Rubella Mixed Titer Performance Panel
Includes Set of 25 aliquots with Anti-Rubella reactivity ranging from negative to strongly positive
Intended Use Evaluating Anti-Rubella tests with characterized samples, to provide comprehensive data for comparative analysis
Matrix Serum and Plasma
Quantity 25 x 0.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.