Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ HCV RNA Linearity Panel

Catalog No. 22554802 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
8 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-802 8 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Follow Universal Precautions: Panel members are positive for HCV RNA and have not been inactivated.
Catalog No. 22-554-802 Supplier SeraCare™ Supplier No. 24100166
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Follow Universal Precautions: Panel members are positive for HCV RNA and have not been inactivated.

Serial dilutions of high titer HCV RNA plasma in HCV RNA negative pooled plasma

  • Allows assessment of linearity through the dynamic range of commercially available and research assays
  • May be used for optimizing, evaluating, monitoring, and troubleshooting these assays
  • May also be used as part of an operator training program, or for competency assessment as part of an ongoing laboratory quality assurance program
  • For research use only; Not for use in diagnostic procedures

Specifications

Content And Storage -70°C
Format Panel
Type HCV RNA Linearity Panel
Includes Set of seven positive members, one negative for HCV RNA
Intended Use For monitoring HCV RNA recovery at defined intervals, identifying consistency over a linear range, verifying lot changes, for operator training program, for operator validation, assessment of linearity over dynamic range of commercially available tests
Quantity 8 x 1.2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.